missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin, MP Biomedicals™

Product Code. 11442701 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
50 mg
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Active against many DNA and RNA viruses. Ribavirin, MP Biomedicals is an antiviral compound that is active against RSV, HIV, HCV and other infectious agents. It is a nucleoside antimetabolite drug that interferes with duplication of viral genetic material

Ribavirin is used as an antiviral agent against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. It inhibits purine (GTP pools) biosynthesis and is a precursor to the di- and tri-phosphates that inhibit viral RNA-dependent RNA polymerases. Its metabolite, ribavirin 5'-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase, but many other mechanisms of action are also supported with experimental evidence. It is also an analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.207
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
Synonym ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
PubChem CID 37542
ChEBI CHEBI:63580
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N

Specifications

Melting Point 173°C (literature)
Loss on Drying 0.04% (Lit.)
Quantity 10 mg
Solubility Information Soluble in water (20mg/mL, clear, colorless solution).
Formula Weight 244.2g/mol
Percent Purity >99%
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.