missing translation for 'onlineSavingsMsg'
Learn More

Ripretinib, MedChemExpress

Product Code. p-200102055
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464618

Brand: Medchem Express HY112306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ripretinib (DCC-2618) is an orally bioavailable, selective KIT and PDGFRA switch-control inhibitor.

Ripretinib (DCC-2618) targets and binds to both wild-type and mutant forms of KIT and PDGFRA specifically at their switch pocket binding sites, thereby preventing the switch from inactive to active conformations of these kinases and inactivating their wild-type and mutant forms. Ripretinib (DCC-2618) also inhibits multiple other kinase targets, such as FLT3 and KDR (or VEGFR-2). DCC-2618 exerts antineoplastic effect and induces apoptosis.

Specifications

Chemical Name or Material Ripretinib
CAS 1442472-39-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H21BrFN5O2
Quantity 10 mg
Synonym DCC-2618
Solubility Information DMSO : 25 mg/mL (48.99 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C2=CC3=C(C=C(NC)N=C3)N(CC)C2=O)=C(Br)C=C1F)NC4=CC=CC=C4
Molecular Weight (g/mol) 510.36
Formula Weight 510.36
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.