Learn More
SI Analytics™ Rod Stirrer
SI Analytics™ Rod Stirrer for use with titration heads TZ 1463 and TZ 1467.
Brand: SI Analytics™ 285215175
This item is not returnable.
View return policy
Description
- Short model
- With NS 14.5
- For titration heads TZ 1463 and TZ 1467
Especificaciones
Short Model | |
Magnetic Stirrer | |
NS 14.5 |