Learn More
Applied Biosystems™ ROI & Background Calibration Kit, 96-well
Shop All Applied Biosystems Products
Click to view available options
Quantity:
2 x 96-well Plates
Unit Size:
Pack of 2
Description
- The 96-Well ROI & Background Calibration Kit is used to maintain your real-time PCR instrument with standard 96-well block.
- Region of interest (ROI) and background calibration should be run after lamp change.
- Please see your instrument user guide for full instructions on use.
Specifications
Specifications
| Content And Storage | Store at -20°C ± 5°C |
| Calibrated Dye | Proprietary Mix |
| For Use With (Equipment) | ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx |
| Product Type | Background Calibration Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Quantity | 2 x 96-well Plates |
| Calibration Type | Spectral Calibration |