missing translation for 'onlineSavingsMsg'
Learn More

Rose Bengal, MP Biomedicals™

Product Code. 11405432
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 632-68-8
Molecular Formula C20H2Cl4I4K2O5
Molecular Weight (g/mol) 1049.845
MDL Number MFCD00005043
InChI Key VQHHOXOLUXRQFQ-UHFFFAOYSA-L
Synonym Acid Red 94, C.I. 45440
PubChem CID 71553
IUPAC Name dipotassium;4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES C1=C2C(=C(C(=C1I)[O-])I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)[O-])I.[K+].[K+]

Specifications

Formula Weight 1017.65
Color Red to Rust
Quantity 5 g
Physical Form Powder
Chemical Name or Material Rose Bengal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.