Learn More
Rotaflo™ Stopcock Spare Parts
Product Code. 12643032
Click to view available options
Type:
Locking Ring
Stopcock Key
Stopcock Spare Parts
Plug Bore:
3 mm
6 mm
10 mm
Unit Size:
Pack of 5
Product Code. 12643032
Brand: Quickfit LR10
Only null left
Add to basket
Add to basket
Replacement locking rings and keys for Quickfit Rotaflo™ stopcocks
Replacement keys and locking rings should be ordered separately
TRUSTED_SUSTAINABILITY
Specifications
Type Stopcock Spare Parts
Plug Bore 10 mm