Learn More
Hirschmann Instruments™ rotarus™ PK 10 Peristaltic Pump Heads
Click to view available options
No. of Channels:
1
Unit Size:
Each
Description
- 3 rolls
- For WS/WT 1.6mm tubes
Specifications
Specifications
| Description | rotarus PK 10 Pump Head |
| No. of Channels | 1 |
| No. of Rollers | 3 |
| Tubing Type | WT 16mm |