Learn More
Ismatec™ Rotary Piston Pump Head
This pump-head is gentle enough for pumping highly concentrated viable cells.
Products 3
Product Code Brand Flow Rate Price Quantity & Availability  
Product Code Brand Flow Rate Price Quantity & Availability  
11943824
View Documents
Ismatec™
ISM791A
0.45 to 3400 mL/min N/A
N/A
11937935
View Documents
Ismatec™
ISM185A
0.002 to 100 mL/min. N/A
N/A
11977785
View Documents
Ismatec™
ISM720A
0002 to 100 mL/min N/A
N/A
Description
Description
  • Coated aluminum pump-head
  • Can be dismantled for cleaning
  • Self-centering tube-track thanks to concave tube-bed and convex rollers, which lengthens the tube-life
  • 3 stainless steel rollers (less pulsation but lower max. flow rate than with 2 rollers)
  • For tubing with 1.6 mm wall thickness
  • 1.5 bar differential pressure