Learn More
Grant Instruments™ Rotor for PCV Series Compact Benchtop Centrifuges
Product Code. p-8014649
Click to view available options
No. of Places:
16
16 (2 microtube strips)
18
Unit Size:
Each
Product Code. 11481028
Brand: Grant Instruments™ PSR16
Fits PCV-2400 Combi-Spin and PVC6400 Multi-spin benchtop centrifuges. Grant Intstruments™ Rotor for PCV Series Compact Benchtop Centrifuges provide versatility with interchangeable rotors for various microtube sizes.
  • PR2-05: Interchangeable centrifuge rotor for 8 x 1.5/2.0mL plus 8 x 0.5mL microtubes (optional)
  • PR2-05-02 : Interchangeable centrifuge rotor for 6 x 1.5/2.0mL plus 6 x 0.2mL microtubes (optional)
  • PSR-16: Interchangeable centrifuge rotor for 2 x 8-well 0.2mL microtube strips (optional)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 2 x 8-well 0.2 mL Microtube Strips
No. of Places 16 (2 microtube strips)