Learn More
Gilac™ Round Basin
Round Basin for food contact. Highly durable and chemical resistant.
Marca: Gilac™ G606721
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Basin fit for food contact
- Highly durable and chemical resistant
- Easy to clean
- Nesting with no suction effect
- Moulded rim makes it easy to grasp
Specifica
Storage Tub | |
White | |
2 gal. | |
Food Contact Certified | |
Nestable | |
4.9 in. | |
16.23 oz. | |
5°F to 140°F |
LDPE | |
Circular | |
7.5 L | |
Food Contact | |
125 mm | |
350 mm | |
0.46 kg | |
-15°C to +60°C |