Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07030094
 View more versions of this product
Product Code. 11933621
  • 615.00 EUR / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Description
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
76 mm
515 mm
1 Ea.
20 L
Beaded rim