Learn More
Gilac™ Round Selective Sorting Dustbin
Click to view available options
Capacity (Metric):
50 L
75 L
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Description
- Round dustbin great as a recycling bin.
- Highly durable.
- Reinforced bottom with anti-wear skirt.
- Reinforced handles to prevent distortion during carrying.
- Optional, choice of two lids.
- External Diameter includes handles.
- Width corresponds to diameter without handles.
Specifications
Specifications
| Capacity (Metric) | 75 L |
| Diameter (Metric) Outer | 670 mm |
| Diameter (English) Outer | 26.4 in. |
| Capacity (English) | 19.8 gal. |
| Color | Yellow |
| Height (Metric) | 485 mm |
| Shape | Round |
| Autoclavable | Not Autoclavable |
| Height (English) | 19.1 in. |
| Material | HDPE |
| Show More |