missing translation for 'onlineSavingsMsg'
Learn More

RP-54745, MedChemExpress

Product Code. 16460496
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16460496

Brand: Medchem Express HY101716

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress RP-54745 is an inhibitor of macrophage stimulation and interleukin-1 production, and a potential antirheumatic compound.

RP-54745 is an inhibitor of macrophage stimulation and interleukin-1 production, and a potential antirheumatic compound.

Specifications

Chemical Name or Material RP-54745
CAS 135330-08-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C13H12ClNOS2
Quantity 20 mg
Solubility Information DMSO : 62.5 mg/mL (209.86 mM; Need ultrasonic)
SMILES O=C1SSC(N2C(C)C3=C(C=CC=C3)CC2)=C1Cl
Molecular Weight (g/mol) 297.82
Formula Weight 297.82
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.