missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14909-1-AP-20UL
This item is not returnable.
View return policy
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S26E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
RPS26 | |
Polyclonal | |
Unconjugated | |
RPS26 | |
40S ribosomal protein S26, ribosomal protein S26, RPS26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27139, 27370, 6231 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62854, P62855, P62856 | |
Rps26 | |
RPS26 Fusion Protein Ag6706 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |