Learn More
Abeba™ Rubber 5100 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather white
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Leather top layer
- Adjustable instep area
- Lining microfiber, respiration absorbing
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC |
| Color | White |
| Liner Material | Microfiber |
| Material | Leather |
| Size | 41 |
| Sole Features | Slip-resistant |
| Sole Material | Rubber |
| Style | 5100 |