Learn More
Abeba™ Rubber 5310 Shoes
ESD dissipative shoe is acid and base resistant.
Brand: Abeba™ 5310/40
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- Black
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Leather top layer
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Leather | |
Slip-resistant | |
5310 |
Black | |
40 | |
Rubber |