Learn More
Abeba™ Rubber 9310 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
Brand:  Abeba™ 9310/42
 View more versions of this product
Product Code. 15603248
  • 69.65 EUR / 1 pair
Estimated Shipment: 03-03-2025
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • Black
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li
Leather
Slip-resistant
9310
Black
42
Rubber