Learn More
Rubidium Chloride, Metal min. 99%., MP Biomedicals
Solutions may be slightly hazy
62.55 EUR - 1000.00 EUR
Chemical Identifiers
CAS | 7791-11-9 |
---|---|
Molecular Formula | ClRb |
Molecular Weight (g/mol) | 120.92 |
MDL Number | MFCD00011187 |
InChI Key | FGDZQCVHDSGLHJ-UHFFFAOYSA-M |
Synonym | rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride |
PubChem CID | 62683 |
ChEBI | CHEBI:78672 |
IUPAC Name | rubidium(1+) chloride |
SMILES | [Cl-].[Rb+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11486202
|
MP Biomedicals
0210282505 |
5 g |
62.55 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11496202
|
MP Biomedicals
0210282510 |
10 g |
104.00 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406212
|
MP Biomedicals
0210282550 |
50 g |
502.00 EUR
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416212
|
MP Biomedicals
0210282580 |
100 g |
1000.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7791-11-9 | |
120.92 | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
62683 | |
rubidium(1+) chloride |
ClRb | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
CHEBI:78672 | |
[Cl-].[Rb+] |
Specifications
7791-11-9 | |
ClRb | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
[Cl-].[Rb+] | |
120.92 | |
CHEBI:78672 | |
Rubidium Chloride |
Metal min. 99%. | |
MFCD00011187 | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
rubidium(1+) chloride | |
62683 | |
120.9 |
Safety and Handling
Recommended Storage : Room Temperature