Learn More
uvex Rubiflex S NB35SF Safety Glove
Shop All Uvex Products
Click to view available options
Size:
8
9
10
11
Description
- Good heat/cold insulation on the palm
- Abrasion and cut resistance
- Ergonomic fit
- High flexibility
- Excellent mechanical characteristics
- Good resistance to many chemicals, acids, alkalis, mineral oils and solvents
- High water vapor absorption due to the cotton lining
- Fully coated
- Sandwich lining on palm, cuff
Specifications
Specifications
Certifications/Compliance | EN: 388 (2 1 2 1 X) |
Color | Green |
Latex Free | Yes |
Material | Cotton, Fleece, NBR |
Product Type | Chemical-resistant Gloves |
Size | 9 |