missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium Red, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Ruthenium Red blocks Ca2+ uptake and release from mitochondria, and Ca2+ release from ryanodine-sensitive intracellular stores. Also blocks cell membrane-located capsaicin-activated cation channels (IC50 = 14 nM) and voltage-sensitive Ca2+ channels to inhibit neurotransmitter release.
Specifications
Specifications
CAS | 11103-72-3 |
Quantity | 100 mg |
Molecular Formula | Cl6H46N14O2Ru3 |
Synonym | 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride |
InChI Key | JQJSTVUROJELSR-UHFFFAOYSA-H |
SMILES | N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] |
IUPAC Name | azane;ruthenium(2+);hexachloride;dihydrate |
Molecular Weight (g/mol) | 790.374 |
PubChem CID | 16218584 |
Formula Weight | 786.35 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction