missing translation for 'onlineSavingsMsg'
Learn More

IKA™ RV 10 digital pro V Double, Complete Package
GREENER_CHOICE

The RV 10 digital pro V double complete package is the complete solution for your rotary evaporator setup with two rotary evaporators.

Brand:  IKA™ 0010011964

 View more versions of this product

Product Code. 17833464

  • 20593.00 EUR / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The RV 10 digital pro V double complete package includes two RV 10 digital V rotary evaporators, HB digital heating bath, vertical glass condenser set, VACSTAR digital vacuum pump, two VC 10 pro vacuum controllers, and RC 2 GREEN basic recirculating chiller.
  • RV 10 digital is a reliable lab companion with an optimized bath volume, efficient condenser unit, and safety features like motorized lift-out function and RS 232 interface for remote control.
  • VACSTAR digital vacuum pump is space-saving and can supply both rotary evaporators, controlled with the help of the two included vacuum controllers.
  • RC 2 GREEN basic recirculating chiller is energy-efficient and suitable for cooling tasks down to -30°C, with demand and speed-controlled compressor operated with natural refrigerant R290.
Specifications

Specifications

440 mm
430 mm
200/240 V
5 to 280 rpm
1600 cm²
3778 W
IP20
2+3 Years Warranty
50/60 Hz
Automatically controlled vacuum/cooling system based on motorized lift with safety-stop function
121.8 kg
500 mm
Ambient to 180°C
RV 10 digital V (2/Cs.), VC 10 pro (2/Cs.), VACSTAR digital, VSE 1, VC 10.400 Stand (2/Cs.), VC 10.300 Check vpcsalve (2/Cs.), RC 2 GREEN basic, RV 10.5005 Set of hoses (2/Cs.), Y hose connector 8 mm (2/Cs.)
Vertical Condenser
2600 W
Distilling Rotary Evaporator
EU
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.