missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-3-methyl-1-butanol, 96%

Product Code. 10627314 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10627314

Brand: Thermo Scientific Acros 181590050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2026-48-4 | C5H13NO | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2026-48-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064296
InChI Key NWYYWIJOWOLJNR-RXMQYKEDSA-N
Synonym l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol
PubChem CID 640993
IUPAC Name (2S)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@H](N)CO

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Yellow
Density 0.9200g/mL
Boiling Point 81.0°C (8.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(NH2)CH2OH
Refractive Index 1.4538 to 1.4558
Quantity 5 mL
Fieser 16,38
Specific Gravity 0.92
Specific Rotation Condition +16.00 (20.00°C c=10,ethanol)
Specific Rotation + 16.00
Formula Weight 103.16
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material (S)-(+)-2-Amino-3-methyl-1-butanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.