Learn More
(S)-(+)-2-Aminohexane, ChiPros 99+%, ee 99+%, Thermo Scientific™
Product Code. 11354206
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(S)-(+)-2-Aminohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water (17g/L at 20°C)
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70492-67-0
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00671626
InChI Key WGBBUURBHXLGFM-UHFFFAOYNA-N
Synonym s-2-aminohexane, 2s-hexan-2-amine, s-2-hexylamine, s-+-2-aminohexane, s-+-2-aminohexane, chipros, ee 99+%, s-2-hexaneamine, pubchem6750, 2s-2-hexanamine, 2-hexanamine, 2s, +-s-2-aminohexane
PubChem CID 6999846
SMILES CCCCC(C)N
Specifications
Melting Point <-20°C
Density 0.755
Boiling Point 116°C
Flash Point −3°C (26°F)
Quantity 5 g
UN Number UN2733
Beilstein 5237601
Sensitivity Air Sensitive
Solubility Information Soluble in water (17g/L at 20°C)
IUPAC Name (2S)-hexan-2-amine
Formula Weight 101.19
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminohexane