missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-(4-cyanophenyl)propionic acid, 95%, Thermo Scientific™

Product Code. 15479555
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 718596-77-1
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD04117832
InChI Key JFPLLJRLBHIJPS-UHFFFAOYNA-N
Synonym s-3-amino-3-4-cyanophenyl propanoic acid, s-3-amino-3-4-cyano-phenyl-propionic acid, 3s-3-amino-3-4-cyanophenyl propanoic acid, s-3-amino-3-4-cyanophenyl propionic acid, h-beta-phe 4-cn-oh, 4-cyano-l-beta-phenylalanine, s-beta-amino-4-cyanohydrocinnamic acid, s-3-amino-4-cyanophenyl propionic acid, s-3-amino-3-4-cyanophenyl propanoicacid, benzenepropanoic acid, b-amino-4-cyano-, bs
PubChem CID 1491714
SMILES NC(CC(O)=O)C1=CC=C(C=C1)C#N

Specifications

IUPAC Name (3S)-3-amino-3-(4-cyanophenyl)propanoic acid
Formula Weight 190.2
Percent Purity 95%
Quantity 250 mg
Sensitivity Air Sensitive
Chemical Name or Material (S)-3-Amino-3-(4-cyanophenyl)propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'