missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Amino-2-(Boc-amino)butyric acid, 97%

Product Code. p-7054328
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15498958

Brand: Thermo Scientific Alfa Aesar H62450.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 25691-37-6 | C9H18N2O4 | 218.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25691-37-6
Molecular Formula C9H18N2O4
Molecular Weight (g/mol) 218.253
MDL Number MFCD00236841
InChI Key MDCPCLPRWLKUIQ-LURJTMIESA-N
Synonym boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
PubChem CID 7019666
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCN)C(=O)O

Specifications

Color White
Quantity 250 mg
Beilstein 4745487
Formula Weight 218.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-4-Amino-2-(Boc-amino)butyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.