Learn More
Heidolph Instruments S-Box Package 1
Product Code. p-7229024 Shop All Heidolph Instruments Products
Click to view available options
Color:
Blue
Green
Orange
Violet
Unit Size:
Each
Product Code. 17848000
Brand: Heidolph Instruments 4000500000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The S-Box Package 1 includes additional accessories to complete an existing S-Biosystem incubation system.
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Compressor Orange
Concentration Range Blue
Content And Storage Violet
Includes 1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers
Input Water Pressure Orange
Installation Blue
Insulation Violet