missing translation for 'onlineSavingsMsg'
Learn More

S-Ethyl thioacetate, 98+%, Thermo Scientific Chemicals

26.93 EUR - 278.00 EUR

Chemical Identifiers

CAS 625-60-5
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00015178
InChI Key APTGPWJUOYMUCE-UHFFFAOYSA-N
Synonym s-ethyl thioacetate, ethanethioic acid, ethyl ester, ethanethioic acid, s-ethyl ester, ethyl thiolacetate, ethyl ethanethioate, ethanethioic acid s-ethyl ester, s-ethyl thiolacetate, acetic acid, thio-, ethyl ester, thioethyl compound, acetic acid, thio-, s-ethyl ester
PubChem CID 61171
IUPAC Name S-ethyl ethanethioate
SMILES CCSC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423444
SDS View Documents
Thermo Scientific Alfa Aesar
A10680.06
5 g
26.93 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
11433444
SDS View Documents
Thermo Scientific Alfa Aesar
A10680.14
25 g
87.33 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
11443444
SDS View Documents
Thermo Scientific Alfa Aesar
A10680.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 210-904-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S-Ethyl thioacetate, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.