Learn More
Thermo Scientific™ Sabouraud Glucose Selective Agar with Gentamicin and Chloramphenicol
Description
Selectively detect and isolate dermatophytes, other fungi and yeasts with Thermo Scientific™ Sabouraud Glucose Agar with Gentamicin and Chloramphenicol. The use of gentamicin and chloramphenicol leads to the following selective effects with only minimal compromises of the growth properties: Chloramphenicol is a broadband antibiotic that suppresses gram-positive and gram-negative bacteria as well as acid-resistant bacilli; however, the growth of Pseudomonas species is only slightly suppressed while gentamicin is particularly effective against P. aeruginosa.
Dermatophytes and fungi grow in typical, characteristic colony shapes on Sabouraud Glucose Agar with Gentamicin and Chloramphenicol. For example, T. rubrum forms 2 to 3cm large, cream-coloured colonies with white spores and pink-colored underside, while A. niger forms approx. 3 to 5cm large, white colonies with black spores and C . albicans forms 2 to 3mm large, yellowish and round colonies. For additional, detailed colony descriptions, please refer to the mycological literature, e.g., Seeliger1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Ivory, transparent |
| Content And Storage | 6°C to 12°C |
| Description | Sabouraud Glucose Selective Agar with Gentamicin and Chloramphenicol |
| Format | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 17g ± 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
For In Vitro Diagnostic Use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.