missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Safe-Lock Tubes
The hinged lid of Eppendorf Safe-Lock Tubes offers outstanding protection against unintentional opening, and completely eliminates any danger of leaks during incubation and storage.
Specifications
Cap Style | Snap cap |
---|---|
Certifications/Compliance | EN 14254:2004 |
Description | Eppendorf Quality™ |
Autoclavable | Autoclavable |
Color | Clear |
Description
- Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- g-Safe™: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30.000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage - Large frosted lid and surface on the side for easy labeling
- Ensured functionality from -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
Specifications
Snap cap | |
Eppendorf Quality™ | |
Clear | |
1000 | |
No |
EN 14254:2004 | |
Autoclavable | |
Polypropylene | |
-86 – 100°C | |
Yes |
Eppendorf Safe-Lock Tubes are single use sample tubes intended as primary vessel in accordance with EN 14254:2004 and for storing human test specimen, with the exception of blood samples for in vitro diagnostics. Eppendorf Safe-Lock Tubes are intended for use by trained and skilled personnel only.