Learn More
Justrite™ Safety Can For Laboratories
Product Code. 12931395
Click to view available options
Capacity (Metric):
19 L
Unit Size:
Each
Product Code. 12931395
Brand: Justrite™ 7150240Z
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
with tap 08540, self closing, controlled dispensing steel, fill spout has self closing pressure relief cap & flame arrester yellow 19L
Specifications
Capacity (Metric) 19 L