missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Safety Canisters
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
12 L
20 L
30 L
60 L
Closure Size:
GL45
S51
S55
S60/61
S60/61 and GL45
S70/71
S90
S95
Includes:
2 openings
Floater
Mechanical Level Control
UN-X approval
UN-X approval; with inner fluorination
UN-Y approval
UN-Y approval, with inner fluorination
Description
- Made of robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
- Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
- Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited under tables or in safety cabinets
- Thread types S55, S60/61, S65, S70/71, S90 and S95 are commonly used in the laboratory, they are all compatible with the SCAT SafetyWasteCaps range for the safe disposal of volatile hazardous substances
- Special versions available with floats for level indication or with internal fluorination for double protection of the contents, both for permeation from outside to inside and for diffusion from inside to outside
The wide range of non-electrically conductive canisters includes a variety of designs for individual use in the laboratory for the disposal of solvents in combination with the SCAT SafetyWasteCaps to increase your laboratory safety.
Specifications
Specifications
Autoclavable | Not Autoclavable |
Capacity (English) | 7.9 gal. |
Capacity (Metric) | 30 L |
Closure Size | S60/61 and GL45 |
Color | White (Translucent) |
Description | Canister 30 L, with 2 openings S60/61 and GL45 PE-HD, dimentions (W x H x D): 290 x 400 x 380 mm ***UN-Y approval invalid*** |
Dimensions (L x W x H) | 290 x 380 x 400 mm |
For Use With (Application) | For All Purposes and Suitable for The S.C.A.T™ SafetySystem |
Includes | 2 openings |
Material | HDPE |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'