Learn More
Brady™ Safety Sign COVID-19 Keep Social Distance
An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.
Brand: Brady™ 306847
1.71 EUR valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Additional Details : Weight : 0.02700kg
Description
- Provides clear, visual communication where it matters
- Highlights the use of appropriate personal protection
- Easily communicates social distancing and hygiene
Specifications
Laminated Polyester | |
Danish | |
Red |
Self-adhesive | |
800 x 80 mm (W x H) |