Learn More
SCAT Safety Waste Caps, with Grounding Connection
GREENER_CHOICE
Product Code. 10262470 Shop All SCAT Europe Products
Click to view available options
Size:
B53
B83
S50
S51
S55
S60/61
S65
S70/71
S90
Includes:
2 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
3 x 2.3/3.2 mm OD
3 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
3 x 2.3/3.2 mm OD, Mechanical Level Control
4 x 2.3/3.2 mm OD
4 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
Unit Size:
Each
Product Code. 10262470
Brand: SCAT 107935
Only null left
Add to basket
Add to basket
Safety Waste Caps with grounding connector to dissipate static charge on the Waste Cap to prevent explosion or fire from sparks.
The SCAT Safety Waste Caps provide maximum safety for your HPLC liquid disposal. Safely dispose of your liquid laboratory waste without contaminating the ambient air by using SafetyWasteCaps with integrated exhaust air filters. These high-performance filters regulate the air pressure balance and prevent the escape of vapors due to the resulting overpressure in the disposal container. The variety of closures, connections and thread sizes allows optimal adaptation to your HPLC situation. Additional versions with grounding connections, level controls or safety funnels or offer every convenience for safe disposal.
  • High quality materials ensure maximum safety for lab and staff
  • Can be individually equipped with level indicators, safety funnels or grounding connections
  • 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
  • Connection port for a SCAT exhaust filter V3.0 on each Safety Waste Cap
Specifications
For Use With (Equipment) Waste Disposal Container
Includes 3 x 2.3/3.2 mm OD
Size S51
For Use With (Application) Waste Disposal System
Type Safety Waste Cap