Learn More
SCAT Safety Waste Set V2.0
Shop All SCAT Europe Products
Click to view available options
Volume (Metric):
10 L Canister
2.5 L Canister
5 L Canister
Includes:
Safety Funnel with Lid 'Arnold'
Safety funnel with lid 'MARCO'
Unit Size:
1 set
Each
Description
- Collect liquid waste safely with the SCAT. safety funnel MARCO V2.0
- Ready to use
- Consists of lid funnel and different canister sizes in a complete set
Specifications
Specifications
| Diameter (Metric) Inner | 140 mm |
| Length (Metric) | 200 mm |
| Includes | Safety Funnel with Lid 'Arnold' |
| Material | HDPE |
| Thread Size | S55 |
| Volume (Metric) | 10 L Canister |
| Type | Waste Funnel |
| Diameter (Metric) Outer | 200 mm |