Learn More
S Murray and Co™ SAMCO™ Dissecting Scissors
Click to view available options
Length (Metric):
112 mm
125 mm
150 mm
Blade Style:
Curved
Straight
missing translation for 'unitSize'
Each
Description
- Stainless Steel dissecting scissors
Specifications
Specifications
| Product Type | Dissecting Scissors |
| Blade Style | Curved |
| Length (Metric) | 112 mm |
| Material | Stainless Steel |