Learn More
Thermo Scientific™ Samco™ Extra Long Transfer Pipettes
GREENER_CHOICE
Product Code. p-4525507
Click to view available options
Volume (Metric):
5.6 mL
6 mL
10.7 mL
23 mL
Sterility:
Non-sterile
Sterile
Packaging Type:
10/Bag, 400/Pk.
20/Bag, 100/Pk., 800/Cs.
20/Bag, 400/Pk., 3200/Cs.
Bulk
Individually Wrapped
Individually Wrapped, White Box
White Box
Quantity:
10/Bag, 400/Pack
10/Bag, 400/Pk.
100/Pk 10Pk/Cs 1000/Cs
100/Pk 8Pk/Cs 800/Cs
100/Pk.
20/Bag, 100/Pk.
20/Bag, 100/Pk., 800/Cs.
20/Bag, 400/Pk.
20/Bag, 400/Pk., 3200/Cs.
400/Pk 8Pk/Cs 3200/Cs
400/Pk 8Pk/Cs 4000/Cs
400/Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 400
Product Code. 15357853
Brand: Thermo Scientific™ 262
Only null left
Add to basket
Edge
Add to basket
Perform safe and easy sampling from large containers using these extra-long transfer pipettes, perfect for research applications.
Perfect for research applications, these Extra-Long Transfer Pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, and autoclaving and prevents cross-contamination. They allow safe and easy sampling from large containers.
  • Excellent for research applications
  • General-purpose pipettes are ideal for routine sampling of liquids
  • Well suited for use with large containers
TRUSTED_SUSTAINABILITY
Specifications
Bulb Draw (Metric) 2.3 mL
Diameter (Metric) Outer 5.0 mm
Material LDPE
Sterility Non-sterile
Drops/mL 25
Graduated No
Length (English) 9 in.
Quantity 400/Pk 8Pk/Cs 4000/Cs
Packaging Type Bulk
Volume (Metric) 5.6 mL
Length (Metric) 22.5 cm
Type Extra Long Transfer Pipet