Learn More
BOLA Sampling Bags
Product Code. 10745982
Click to view available options
Capacity (Metric):
1 L
2 L
5 L
10 L
Unit Size:
Each
Product Code. 10745982
Brand: BOLA A22374
Only null left
Add to basket
Add to basket
Sampling Bags PVF, 2 connectors
  • Transparent
  • Non-porous
  • Without plasticisers
  • With 2 exchangeable ports made of PTFE.
Specifications
Capacity (Metric) 10 L
Color Transparent
Length (Metric) 350 mm
Width (Metric) 500 mm