missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Santonin, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 481-06-1 |
Molecular Formula | C15H18O3 |
MDL Number | MFCD00135865 |
Quantity | 10 g |
Synonym | santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum |
InChI Key | XJHDMGJURBVLLE-BOCCBSBMSA-N |
SMILES | C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O |
IUPAC Name | (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione |
Molecular Weight (g/mol) | 246.31 |
PubChem CID | 221071 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction