Learn More
Saridegib, MedChemExpress
MedChemExpress Saridegib is a potent and specific inhibitor of Smoothened (Smo), a key signaling transmembrane protein in the Hedgehog (Hh) pathway.
Brand: Medchem Express HY-16587
This item is not returnable.
View return policy
Description
Saridegib is a potent and specific inhibitor of Smoothened (Smo), a key signaling transmembrane protein in the Hedgehog (Hh) pathway.Specifications
Saridegib | |
White | |
C29H48N2O3S | |
IPI-926 Patidegib | |
504.77 | |
99.0% | |
Solid |
1037210-93-7 | |
Research | |
10 mg | |
CC1=C(C[C@@]2([H])[C@@]3([H])CC[C@@]4([H])[C@]2(C)CC[C@@H](NS(C)(=O)=O)C4)[C@@]3([H])CC[C@@]([C@@H]5C)(C1)O[C@@]6([H])[C@@]5([H])NC[C@@H](C)C6 | |
504.77 | |
Research | |
Cancer-programmed cell death |