Learn More
SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI3A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA814466
Encompass_Preferred
 View more versions of this product
Product Code. 17599214
  • 540.87 EUR / 100µL
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA, Western Blot
Specifications
Specifications
SARS-CoV-2 Spike Protein
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Mouse
Protein A/G
RUO
Virus
Antibody
IgG2b
ELISA, Western Blot
OTI3A2
Unconjugated
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli.
100 μL
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions