missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Nutrient Pad Sets (NPS) for Microsart™ e.motion Dispenser

Product Code. 15942490 Shop All Sartorius Products
Click to view available options
Media Type:
Azide
Cetrimide
Endo
Orange Serum
Schaufus Pottinger
Schaufus Pottinger (m-Green Yeast and Mold)
Standard TTC
TGE
Tergitol TTC
Wort
Diameter (Metric) Outside:
4.7 cm
5 cm
Pore Size:
0.45μm
0.65μm
0.8μm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15942490

Brand: Sartorius 1406250RDN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are sterile, dehydrated culture media designed for use with membrane filter methods. Each Nutrient Pad Set includes the appropriate membrane filters (in 47 or 50mm diameters), which are also sterile and individually packaged. Just moisten the Nutrient Pads with sterile and demineralized (or distilled) water and they are immediately ready to use. They comply with international regulations and recommendations including, but not limited to: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA.

  • Trouble-free storage up to 24 months
  • Economical -- time and cost savings
  • Easy handling
  • Validation guide available
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN and ISO standards
Color Green
Form Dehydrated
Manufacturing Origin Germany
Media Type Orange Serum
Product Type Orange Serum
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) Acid-tolerant microorganisms
Grid Color Dark Green
Packaging Type Individually Packaged
Pore Size 0.45μm
Diameter (English) Outside 1.96 in.
Diameter (Metric) Outside 5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.