missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 747651A dihydrochloride
GREENER_CHOICE

Product Code. 30001551
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30001551

Brand: Tocris Bioscience™ 4630/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent MSK1 inhibitor; also inhibits other AGC group kinases

Potent, ATP-competitive mitogen- and stress-activated kinase 1 (MSK1) inhibitor (IC50 = 11 nM in an in vitro kinase assay); targets the N-terminal kinase domain. Inhibits MSK1, MSK2, PKA, PKB, RSK and p70S6K activity in cells. Blocks IL-10 production in macrophages.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1781882-72-1
Quantity 10 mg
Molecular Formula C16H24Cl2N8O
Formula Weight 415.32
Purity >98%
Molecular Weight (g/mol) 415.323
Chemical Name or Material SB 747651A dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.