missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SC144, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
SC144 is a first-in-class, orally active gp130 (IL6-beta) inhibitor. SC144 binds gp130, induces gp130 phosphorylation (S782) and deglycosylation, abrogates Stat3 phosphorylation and nuclear translocation, and further inhibits the expression of downstream target genes. SC144 shows potent inhibition of gp130 ligand-triggered signaling. SC144 induces apoptosis in human ovarian cancer cells.
Specifications
Specifications
| Chemical Name or Material | SC144 |
| CAS | 895158-95-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C16H11FN6O |
| Quantity | 100 mg |
| Solubility Information | DMSO : 16.67 mg/mL (51.72 mM; Need ultrasonic) |
| SMILES | O=C(C1=NC=CN=C1)NNC2=NC3=C(N4C2=CC=C4)C=CC(F)=C3 |
| Molecular Weight (g/mol) | 322.3 |
| Formula Weight | 322.3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction