missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Schaedler Anaerobe Agar with Sheep Blood, Haemin, Vitamin K1
Click to view available options
Description:
Schaedler Anaerobe Agar with Sheep Blood with Haemin and Vitamin K1
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Grow and isolate obligate and facultative anaerobic organisms, such as the fastidious anaerobic bacteria Bacteroides, Prevotella and Porphyromonas, using Thermo Scientific™ Schaedler Anaerobe Agar with Sheep Blood, Haemin and Vitamin K1.
Schaedler anaerobe agar is free from thioglycollate and is used to create selective conditions under which the required, delicate and more nutritionally exacting microorganisms of the intestinal tract would develop, despite the presence of antagonistic organisms. Normally such fastidious microorganisms would be swamped by the growth of enterococci, coliform bacilli and other Gram-negative bacilli.
Schaedler Anaerobe Agar contains cysteine hydrochloride and glucose, as reducing substances, with the advantage that cysteine inhibits the growth of Escherichia coli. Schaedler Anaerobe Agar has been shown to be a suitable alternative to blood agar for the enumeration of clostridia and has been used for the examination of food, waste products and ditch water. Studies have shown the necessity for strict anaerobic conditions for the successful recovery of obligate anaerobes when using this medium without the addition of blood.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Schaedler Anaerobe Agar contains cysteine hydrochloride and glucose, as reducing substances, with the advantage that cysteine inhibits the growth of Escherichia coli. Schaedler Anaerobe Agar has been shown to be a suitable alternative to blood agar for the enumeration of clostridia and has been used for the examination of food, waste products and ditch water. Studies have shown the necessity for strict anaerobic conditions for the successful recovery of obligate anaerobes when using this medium without the addition of blood.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Signal red, opaque |
| Content And Storage | 2° to 12°C, in the dark |
| Description | Schaedler Anaerobe Agar with Sheep Blood with Haemin and Vitamin K1 |
| Format | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 17g ± 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction