Learn More
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
- Cylindrical, Schlenk-style flask
- Includes glass tap and cone
- Available in different capacities
- Available with different joint sizes
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | Schlenk lines and vacuum manifolds |
| Includes | Glass tap and cone |
| Product Type | Cylindrical Schlenk flask with tap and cone |
| Shape | Cylindrical |
| Socket Joint | Joint size: 19 |
| Bottom Shape | Cylindrical |