Learn More
Lenz Laborglasintrumente™ Schlenk Tubes with PTFE Needle-Valve Stopcock
Product Code. p-7161785
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
Type:
Cone NS 14/23
Socket NS 14/23
Unit Size:
Each
Product Code. 11933421
Brand: Lenz Laborglasintrumente™ 03370813
Nitrogen tubes (Schlenk tubes) with conical ground joint and PTFE needle-valve stopcock
  • With conical ground joint and PTFE needle-valve stopcock
  • Material: DURAN™
Specifications
Capacity (Metric) 10 mL
Material Duran Tubing
Type Cone NS 14/23
Pore Size Valve bore 2.5 mm
Quantity 1