Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
Brand:  SI Analytics™ 285130400
 View more versions of this product
Product Code. 10211503
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sulfide
Ag-S 60
0°C to 80°C
0.003 to 32,000 mg/L
2 to 12
Selective
Electrode