Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
Brand: SI Analytics™ 285130370
This item is not returnable.
View return policy
Description
- ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
- Also measure pH and temperature
Specifications
Potassium | |
K 60 | |
0°C to 40°C | |
0.04 to 39,000 mg/L |
2 to 12 | |
Selective | |
Electrode |