Learn More
DWK Life Sciences Screw Cap closed, with seal for safety standard ground sockets
For safe sealing of DURAN™ threaded sleeves after use.
Brand:  DWK Life Sciences 286214408
 View more versions of this product
Product Code. 15804608
  • 49.10 EUR / Pack of 10
Estimated Shipment: 09-04-2025
Description
With closed screw caps, devices made of DURAN™ safety grinding can be securely closed after use. The PTFE-coated silicone seal provides maximum chemical resistance.
  • Secure closure of the safety ground joints prevents unintentional opening of the ground joint
  • PTFE-coated silicone gasket for maximum chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Suitable for sealing security grinding
NS 24/29