missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHA Mouse anti-Human, Mouse, Rat, Clone: 5E10G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66588-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.Specifications
SDHA | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31040, Q8K2B3, Q920L2 | |
Sdha | |
SDHA Fusion Protein Ag7023 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5E10G12 | |
Unconjugated | |
SDHA | |
FP, SDH2, SDHA, SDHF | |
Mouse | |
Protein G | |
RUO | |
157074, 6389, 66945 | |
-20°C | |
Liquid |